Atomax™ Hollow Cathode Lamps, PerkinElmer
Supplier: PERKINELMER SCIENTIFIC CANADA ULC
Certificates
About this item
Single-element hollow cathode lamps are crafted to the highest standards.
- Single-element design
- High-quality construction
- Near universal compatibility with atomic absorption instruments
Fit most commercially available atomic absorption instruments that require 37mm (11/2") diameter lamps.
Ordering information: Manufacturer's warranty for 5000 milliampere-hours per lamp; contact your Avantor representative for more information.
Customers Who Bought This Also Bought