DuPont™ ProShield® 30 Boot Covers
Supplier: DuPont
Certificates
About this item
ProShield® 30 shoe and boot covers offer exceptional skid resistance.
- Serged seams
- Elastic at ankles
- Skid-resistant
ProShield® 30 is made from spun bonded polypropylene with a polyethylene coating. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic at ankles ensures a secure fit.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Specifications
- Catalog No:
- CA80077-260
- CA89129-708
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory