About this item
Cold sterilant is a peracetic acid solution for use in a variety of medical, pharmaceutical, and industrial applications
Cold sterilant is a peracetic acid solution for use in a variety of medical, pharmaceutical, and industrial applications
It also provides protection for reverse osmosis membranes and associated distribution systems against short- and long-term bacterial problems. Solution provides 6 log reduction of bacteria population at a 100X dilution (1%) within a 36-minute soak time. Does not produce toxic aldehyde vapors. Biodegradable and easily disposable. Compatible with thin film composite membranes.