About this item
Safe, durable water baths feature an integrated digital controller keypad with an LED that displays the temperature to 0.1°C
The tank is stainless steel with no welded seams and an exterior housing that features an electrostatic powder-paint finish and inner drip plates to prevent condensation. The internal chamber's corners are rounded for easy cleaning. A stainless steel hinged gable cover can be fixed in open position or removed completely to optimize bench space. The maximum temperature indicated can be reached only with the cover in use. Stainless steel racks secure vessels within bath, and have polymer coated handles for safe contact.
An embedded holder protects internal electronics, and a temperature cut-off function ensures samples will not be harmed if the temperature exceeds the desired point. The heating elements are restricted to the area below the bath vessel, ensuring homogenous temperature allocation and no exposed heating coils. The heating systems also make it possible to operate the bath down to a minimum water level of 20mm from the bottom of the tank, creating more space in the bath for large vessels
Operating temperature: 5...40 °C
Specifications
- Catalog No:
- 98000-126
- 98000-136
- 98000-112
- 98000-118
- 98000-122
- 98000-120
- Temperature range:
- Temperature stability:
- Temperature set point resolution:
- Electrical: