Propyl-4-hydroxybenzoate 98.0-102.0%, GenAR® NF, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Propyl 4-hydroxybenzoate,Propyl parahydroxybenzoate,Propylparaben
Formula:C₁₀H₁₂O₃
MW:180.2 g/mol
Boiling Pt:292 °C (1013 hPa)
Melting Pt:95 °C
Density:1.28 g/cm³ (20 °C)
MDL Number:MFCD00002354
CAS Number:94-13-3
Merck Index:12,08051
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-102,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 292 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 95 °C
:Flash Pt:
:Flash Pt:
:M.W. 180.2 g/mol
EINECS:202-307-7
:GHS07
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets J.P. Chemical Specifications:
- GMP Manufactured Product:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets N.F. Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Assay (C₁₀H₁₂O₃):98.0 - 102.0 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Color of Solution:Passes Test
- NF - Acidity:Passes Test
- NF - Related Substances - p-Hydroxybenzoic Acid:≤0.5 %
- NF - Related Substances - Unspecified Impurities (each):≤0.5 %
- NF - Related Substances - Total Impurities:≤1.0 %
- NF - Residue on Ignition:≤0.1 %
- EP/BP - Assay (C₁₀H₁₂O₃):98.0 - 102.0 %
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Acidity:Passes Test
- EP/BP - Related Substances - Impurity A:≤0.5 %
- EP/BP - Related Substances - Unspecified Impurities (each):≤0.5 %
- EP/BP - Related Substances - Total Impurities:≤1.0 %
- EP/BP - Sulfated Ash:≤0.1 %
- JP - Assay:98.0 - 102.0
- JP - Identification:Passes Test
- JP - Melting Point:96 - 99 °C
- JP - Acidity:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Heavy Metals:≤20 ppm
- JP-RelatedSubstances- p-Hydroxybenzoic Acid:≤0.5 %
- JP-Related Substances-Unspecified Impurities (each):≤0.5 %
- JP-Related Substances-Total Impurities:≤1.0 %
- JP - Residue on Ignition:≤0.10 %
- Only the Class 3 solvent 1-propanol is present. The concentration is:
- controlled below 0.5%.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
Related Information
Certificates
Search by Lot Number7624 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR7624
SDS
spec_JT7624-06 spec_MK762418
Product specification
Customers Who Bought This Also Bought