4-Methyl-2-pentanone ≥98.5% (by GC, corrected for water content), CMOS for microelectronic, J.T.Baker®
Catalog #: CAJT9405-3
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 108-10-1
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:8 pt.
Specifications
Specification Test Results
- For Microelectronic Use:
- Appearance:Passes Test
- Assay (CH₃COCH₂CH(CH₃)₂) (by GC, corrected for water):≥98.5 %
- Color (APHA):≤15
- Density (g/mL) at 25°C:0.794 - 0.798
- Residue after Evaporation:≤0.0050 %
- Acidity (as CH₃COOH):≤0.01 %
- Water (H₂O)(by Karl Fischer titrn):≤0.1 %
- Particle Count - 0.5 µm and greater:≤130 par/ml
- Particle Count - 1.0 µm and greater:≤10 par/ml
- Boiling Range (Initial):≥114.0 °C
- Boiling Range (Dry Point):≤117.0 °C
- J.T.Baker®, 4-Methyl-2-pentanone ≥98.5% (by GC, corrected for water content), CMOS for microelectronic:
Specification Test Results
About this item
Synonyms: Methyl iso-Butyl Ketone,Isobutyl Methyl Ketone
Boiling Pt:117.4 °C (1013 hPa)
Melting Pt:–84 °C
Density:0.801 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5
:Management of Change (MOC) category = R
Flash Pt:16 °C
:Boiling Pt:
:Boiling Pt: 117.4 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -84 °C|Melting Pt: –84 °C
:Flash Pt:
:Flash Pt: 16 °C
:M.W. 100.16 g/mol
Merck Index:13,05227
Formula:(CH₃)₂CHCH₂COCH₃
MW:100.16 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00008938
CAS Number:108-10-1
EINECS:203-550-1
UN:1245
:3
:II
:GHS02
:GHS07
:GHS09
Customers Who Bought This Also Bought