Methanol ≥99.9% (by GC), CMOS for the electronics industry, J.T.Baker®
Catalog #: JT9073-5
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 67-56-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle, pressure bottom
- Size:4 L
- Supplier No.:9073-05
Specifications
Specification Test Results
- For Microelectronic Use:
- Assay (CH₃OH) (by GC):≥99.9 %
- Color (APHA):≤10
- Acidity (µeq/g):≤0.3
- Alkalinity (µeq/g):≤0.1
- Heavy Metals (as Pb):≤100.0 ppb
- Residue after Evaporation:≤5 ppm
- Water (H₂O)(by Karl Fischer titrn):≤0.05 %
- Solubility in H₂O:Passes Test
- Chloride (Cl):≤0.2 ppm
- Phosphate (PO₄):≤0.3 ppm
- Sulfate (SO₄):≤0.5 ppm
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Trace Impurities - Arsenic (As):≤5.0 ppb
- Trace Impurities - Antimony (Sb):≤5.0 ppb
- Trace Impurities - Barium (Ba):≤20.0 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤20.0 ppb
- Trace Impurities - Calcium (Ca):≤50.0 ppb
- Trace Impurities - Chromium (Cr):≤20.0 ppb
- Trace Impurities - Cobalt (Co):≤20 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gallium (Ga):≤50 ppb
- Trace Impurities - Germanium (Ge):≤50.0 ppb
- Trace Impurities - Gold (Au):≤20 ppb
- Trace Impurities - Iron (Fe):≤50.0 ppb
- Trace Impurities - Lead (Pb):≤50 ppb
- Trace Impurities - Lithium (Li):≤50.0 ppb
- Trace Impurities - Magnesium (Mg):≤50.0 ppb
- Trace Impurities - Manganese (Mn):≤10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤300.0 ppb
- Trace Impurities - Nickel (Ni):≤10.0 ppb
- Trace Impurities - Potassium (K):≤50 ppb
- Trace Impurities - Silicon (Si):≤50 ppb
- Trace Impurities - Silver (Ag):≤20.0 ppb
- Trace Impurities - Sodium (Na):≤50.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tin (Sn):≤50 ppb
- Trace Impurities - Titanium (Ti):≤20 ppb
- Trace Impurities - Vanadium (V):≤300.0 ppb
- Trace Impurities - Zinc (Zn):≤50 ppb
- Trace Impurities - Zirconium (Zr):≤300.0 ppb
- Particle Count - 0.5 µm and greater:≤100 par/ml
- Particle Count - 1.0 µm and greater:≤8 par/ml
- J.T.Baker®, Methanol ≥99.9% (by GC), CMOS for the electronics industry:
Specification Test Results
About this item
Synonyms: Hydroxy methane,Carbinol,Wood alcohol
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
:Management of Change (MOC) category = R
Flash Pt:11 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 64.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -98 °C|Melting Pt: –98 °C
:Flash Pt:
:Flash Pt: 11 °C (closed cup)
:M.W. 32.04 g/mol
Merck Index:13,05984
Formula:CH₃OH
MW:32.04 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00004595
CAS Number:67-56-1
EINECS:200-659-6
UN:1230
REACH:01-2119433307-44
:3
:II
:11 °C
:GHS02
:GHS06
:GHS08
:M.O.C. Category R
Customers Who Bought This Also Bought