L(+)-Glutamine 98.5-101.5% (dry basis) USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Gln,L-Glutamine,H-Gln-OH,(S)-(+)-Glutamine,(S)-2,5-Diamino-5-oxopentanoic acid,Levoglutamide
Formula:C₅H₁₀N₂O₃
MW:146.15 g/mol
Boiling Pt:355 °C (1013 hPa)
Melting Pt:185…186 °C
Density:1.5326 g/cm³ (130 °C)
MDL Number:MFCD00008044
CAS Number:56-85-9
Merck Index:13,04484
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5-101,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 355 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 185 to 186 °C|Melting Pt: 185…186 °C
:Flash Pt:
:Flash Pt:
:M.W. 146.15 g/mol
:L(+)-Glutamine
Storage Temperature:Ambient
EINECS:200-292-1
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets U.S.P Requirements:
- Meets J.P. Chemical Specifications:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (C₅H₁₀N₂O₃) (dried basis):98.5 - 101.5 %
- USP - Identification:Passes Test
- USP - Specific Rotation [Å]²º^D (+):6.3 - 7.3 Degree
- USP - Loss on Drying at 105°C:≤ 0.3 %
- USP - Residue after Ignition:≤ 0.3 %
- USP - Chloride (Cl):≤ 0.05 %
- USP - Sulfate (SO₄):≤ 0.03 %
- USP - Iron (Fe):≤ 0.003 %
- USP - Related Compounds:0.5% max
- JP - Assay (C₅H₁₀N₂O₃) (dried basis):99.0 - 101.0 %
- JP - Identification:Passes Test
- JP - Specific Rotation [Å]²º^D (+):6.3 - 7.3 Degree
- JP - Loss on Drying at 105°C:≤ 0.30 %
- JP - Residue after Ignition:≤ 0.1 %
- JP - Chloride (Cl):≤ 0.021 %
- JP - Sulfate (SO₄):≤ 0.028 %
- JP - Iron (Fe):≤ 10 ppm
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Related substances:Passes Test
- JP - pH:4.5 - 6.0
- JP - Clarity and Color of Solution:Passes Test
- JP - Ammonium:≤ 0.1 %
- Solubility of 3% Solution at 30°C:Passes Test
- Solubility of 3.5% Solution at 30°C:Passes Test
- Identification (by IR):Passes Test
- Endotoxin Concentration (EU/mg):≤ 0.06
- Endotoxin Concentration, IU/mg:Passes Test
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP (232), EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, L(+)-Glutamine 98.5-101.5% (dry basis) USP, Multi-Compendial:
Specification Test Results
Related Information
Certificates
Search by Lot Number2078 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
JTBaker_2078_Datasheet.pdf
Datasheet
spec_JT2078-6 spec_JT2078-7 spec_JT2078-9
Product specification
Customers Who Bought This Also Bought