Sample Containers with Screw Caps, Polypropylene, BrandTech
Supplier: Brandtech
Certificates
About this item
Translucent polypropylene containers with screw caps
- Chemically resistant polypropylene
- Easy to clean
- Screw on caps for security
Maximum operating temperature: 125 °C (225 °F). Autoclavable.
Customers Who Bought This Also Bought