Barnstead™ B-pure™ Filters, Thermo Scientific
Supplier: Thermo Fisher Scientific
CE Compliant
Certificates
About this item
Filters for pre- and post-filtration of water and are compatible with B-pure™ ½ size filter holders.
- Features a quarter-turn quick-release canister for easy replacement
- Natural polypropylene construction eliminates recontamination
Specifications
- Catalog No:
- 26302-444
- 26302-445
- 26302-440
- 26302-441
- 26302-442
- 26302-443
- Inlet/outlet connection:
- 12.7 mm (1/2") NPTF
- 12.7 mm (1/2") NPTF
- 12.7 mm (1/2") NPTF
- 12.7 mm (1/2") NPTF
- 12.7 mm (1/2") NPTF
- 12.7 mm (1/2") NPTF
- Inlet temp. range:
- 4...49°C (40...120°F)
- 4...49°C (40...120°F)
- 4...49°C (40...120°F)
- 4...49°C (40...120°F)
- 4...49°C (40...120°F)
- 4...49°C (40...120°F)
- Max. inlet pressure:
- 100 psig
- 100 psig
- 100 psig
- 100 psig
- 100 psig
- 100 psig