Order Entry
ContactUsLinkComponent
Gill-Style Sample Cups, Independent Can company
Gill-Style Sample Cups, Independent Can company
Catalog #: 56778-128
Supplier:  United State Can
Gill-Style Sample Cups
Gill-Style Sample Cups, Independent Can company
Catalog #: 56778-128
Supplier:  United State Can

Specifications

  • Capacity:
    177 ml (6 oz.)
  • Cat. no.:
    56778-128
  • Supplier No.:
    36000061

Specifications

About this item

Asphalt sample cups are useful for ointments, waxes, other solids, and small parts

Made of tinplate, seamless, with slip-on cover.

Customers Who Bought This Also Bought