About this item
PP, sterile, with or without PE lid.
Polyethylene lid is frosted for easy marking, and fits both container sizes.
- Molded-in graduations for easy measurement
- Inert and chemically resistant to commonly used laboratory reagents at room temperature
PP, sterile, with or without PE lid.
Convenient, single-use containers for collection, transport and storage of a wide variety of specimens.
Polyethylene lid is frosted for easy marking, and fits both container sizes.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Certificates
Search by Lot Number15704-029_06719065 15704-029_10019068 15704-029_21219017 15704-029_23819036 15704-051_11219093
Conformity declaration
Corning New Lab Startup Program
Promotion Material
Corning® Your Favorites are Back in Stock