Corning® Disposable Containers, Polypropylene, Sterile, Corning
Supplier: Corning
Certificates
About this item
PP Container, transparent, with snap-on HDPE lid.
- Graduated in both millilitres and ounces
- Certified non pyrogenic
- sterile
PP Container, transparent, with snap-on HDPE lid.
Serves as a beaker or storage container.
Specifications
- Catalog No:
- 15705-305
- 15705-270
- 15705-338
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
Specifications
Related Information
Certificates
Search by Lot Number15705-270_17223023 15705-305_33322011
Conformity declaration
Corning New Lab Startup Program
Promotion Material
Corning® Your Favorites are Back in Stock
Product Literature
Customers Who Bought This Also Bought