Paint and Ink Safety Storage Cabinets, Eagle Manufacturing
Supplier: Eagle Manufacturing
Certificates
About this item
Cabinets are constructed of tough, double-walled 18-gauge steel with 3.8 cm (1½") air space between walls.
- Enamel finish
- Sliding door models are self-closing with fusible link
- Both upper and lower vents have a fire baffle
- Shelf depth for 454.2 L (120 gal.) models is 71.1 cm (28")
- Shelf depth on 151.4 L (40 gal.) and 227.1 L (60 gal.) models is 37.4 cm (14¾")
Complete with grounding attachments, three-point key lock, and 5 cm (2") raised leakproof door sills. Warning sign 'Flammable–Keep Fire Away' appears in three languages (English, Spanish, French).
Certifications: Conforms to OSHA flammable liquid storage requirements and NFPA Code 30. Self-closing models meet UFC 79.
Customers Who Bought This Also Bought