To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:1 ml
- Cat. no.:RK32024
- Supplier No.:32024
Specifications
About this item
Mixture is packaged in methanol at a concentration of 200 µg/mL in a 1 mL ampoule.
- Mixture contains the compounds Alachlor, Aldrin, Atrazine, [gamma]-BHC (Lindane), [alpha]-Chlordane, [gamma]-Chlordane, Dieldrin, Endrin, Heptachlor, Heptachlor Epoxide B, Hexachlorobenzene, Hexachlorocyclopentadiene, Methoxychlor, cis-Nonachlor, trans-Nonachlor, and Simazine
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).