Specifications
- Description:UST o-Terphenyl Diesel Internal Standard
- Cat. no.:RK31066
- Supplier No.:31066
Specifications
About this item
Standards are packaged in acetone at a concentration of 2000 µg/mL in a 1 mL ampoule.
- 2000 μg/mL in acetone, 1 mL/ampoule
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).