Specifications
- Description:UST 5-[alpha]-Androstane Diesel Internal Standard
- Cat. no.:RK31065
- Supplier No.:31065
Specifications
About this item
Standards are packaged in methylene chloride at a concentration of 2000 µg/mL in a 1 mL ampoule.
- 2000 μg/mL in methylene chloride, 1 mL/ampoule
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).