Specifications
- Concentration:7500 µg/ml
- Description:Acid Matrix Spike Mix
- Size:5 ml
- Cat. no.:RK31085
- Supplier No.:31085
Specifications
About this item
Standards are packaged in methanol at the stated concentration in an ampoule.
- Mixture contains 4-Chloro-3-methylphenol, 2-Chlorophenol, 4-Nitrophenol, Pentachlorophenol, and Phenol
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).