UST Diesel Surrogate Standards, Restek
Supplier: Restek
Certificates
About this item
Standards are packaged in methylene chloride at a concentration of 10000 µg/mL in a 1 mL ampoule.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Delivery: Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.