Stripwell™ 96-Well Polystyrene Low Volume Microplates, Corning®
Supplier: Corning
Certificates
About this item
Corning® Stripwell™ microplates may be used in a variety of laboratory assays. The flat bottom strips are designed to easily break apart and are pre-assembled in an ‘egg-crate’ style strip holder that allows everyone well to be positioned back into the microplate once broken.
- Stripwell microplates have 96-well flat bottom polystyrene format
- Low volume and standard Stripwell microplates have well volumes of 190 µl and 360 µl, respectively
- Strips 1×8 are designed to fit only one way into the strip holder, eliminating the chance of misorientation
Specifications
- Catalog No:
- 46610-186
- 46610-190
- 46610-198
- 46610-192
- 46610-194
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk