VWR® Spinstar® Magnetic Stir Bars
Supplier: VWR International
Certificates
About this item
Spinstar® magnetic stirring bars create a deep mixing vortex at relatively slow speeds
- Star shape is designed to fit the inside diameter of most commonly-used beakers
- Perfect for applications requiring slow, thorough mixing
- Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclave safe
Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low friction coating; durable and inert.