VWR® Spinbar® Polygon Stir Bars, Assorted
Catalog #: 58948-072
Supplier: VWR International
Specifications
- Coating:PTFE
- Description:Stir Bars
- Cat. no.:58948-072
- Shape:Polygon
Specifications
About this item
Assortment of 18 PTFE resin-coated, polygon stir bars contained in a plastic, compartmentalized Spinbox™ with indexed cover.
- PTFE resin coating
- Alnico magnet
- Polygon shape has an interrupted profile for added turbulence
This assortment of stir bars is an economical choice in a durable plastic case.
Ordering information: Includes stir bars in seven sizes: 20x7mm (25/32x9/32"), 25x8mm (31/32x5/16"), 30x8mm (13/16x5/16"), 40x8mm (19/16x5/16"), 50x8mm (131/32x5/16"), 60x8mm (23/8x5/16"), and 80x10mm (35/32x13/32").
Customers Who Bought This Also Bought