About this item
Large volume magnetic stirrers are designed for large capacity applications
Powerful rare earth magnets automatically pull stir bars into position and stay coupled under the most demanding stirring conditions. The rugged ABS/polycarbonate housing and heavy-duty motor offer exceptional magnetic force and provide extended reliability. Stirrers feature non-skid feet to prevent the unit from moving during operation. The MegaMag Genie™ stirrers feature simple analog controls that allow user to easily change the desired speed. The digital MegaMag Genie™ stirrers feature true speed readout with feedback control that is adjustable in 10rpm increments. Digital unit also includes an adjustable timer. The analog and digital MegaMag Genie™ stirrers can stir up to 38L (10gal.)
Specifications
- Catalog No:
- 89037-472
- 89037-468
- Weight:
- 3 kg (6.6 lbs.)
- 3 kg (6.6 lbs.)
- Overall dimensions:
- 13H×30.5W×31D cm (51/8×12×127/32")
- 13H×30.5W×31D cm (51/8×12×127/32")