ContactUsLinkComponent
Nalgene® Stirring Rod, Polypropylene, Thermo Scientific
Nalgene® Stirring Rod, Polypropylene, Thermo Scientific
Catalog #: 59062-097
Nalgene Stirring Rod, Polypropylene, Thermo Scientific
Nalgene® Stirring Rod, Polypropylene, Thermo Scientific
Catalog #: 59062-097
Specifications
  • Description:
    Polypropylene Stirring Rod
  • Cat. no.:
    59062-097
  • Supplier No.:
    6169-0010
Specifications
About this item
Versatile stirring rod with a flat, flexible, 15.9x15.9mm (5/8x5/8") paddle at one end, and a triangular paddle at other end
Length: 247.7mm (93/4"). Combines functions of stirring rod and rubber policeman.
Customers Who Bought This Also Bought