JS-4.0 Swinging Bucket Rotor, Beckman Coulter®
Catalog #: CABK339086
Supplier: Beckman Coulter
Specifications
- Description:JS-4.0 Rotor
- Max. RCF:4050 g
- Max. speed:4000 rpm
- Cat. no.:CABK339086
- Supplier No.:339086
Specifications
About this item
This four-place, aluminum swinging bucket rotor is for use with Avanti™ J-26 XP and XPIBK393124 series and J6-MIBK360291 centrifuge.
- Ideal for Rapid Sedimentation of Protein Precipitates, Large Particles, Cells, and Cell Debris
- Rotor Capacity: Four Liters or Four Blood Bags
- Interchangeable with Microplate Carriers
- Material: Aluminum
- Four-Place Rotors
This rotor is ideal for rapid sedimentation of protein precipitates, large particles, cells, and cell debris
Rotor capacity is 4L or four blood bags. The buckets are interchangeable with microplate carriers (sold separately); each carrier holds three microtiter plates.
It's suitable for binding studies and separating serum from whole blood.
Ordering information: Aeroseal™ cover for round buckets features an O-ring seal for added aerosol protection. For Multi-Disc™ adapters, see BK339100 series.
Customers Who Bought This Also Bought