Cupsink and Turret Kit for Basic™ Laboratory Hoods, Labconco®
Catalog #: 15202-652
Supplier: Labconco
Product available on GSA Advantage®
Specifications
- Description:Cupsink and Turret Kit
- Cat. no.:15202-652
- Supplier No.:4852500
Specifications
About this item
Kit includes a 7.6x15.2cm (3x6") polypropylene cupsink and chrome turret
The cupsink and turret mount on the right side wall. Plumbing pipe is not included.
Customers Who Bought This Also Bought