Order Entry
ContactUsLinkComponent
VWR® Spinbar® Cuvette Stir Bar
VWR® Spinbar® Cuvette Stir Bar
Catalog #: 58949-030
Supplier:  VWR International
VWR* Cuvette Spinbar* Stir Bar
VWR® Spinbar® Cuvette Stir Bar
Catalog #: 58949-030
Supplier:  VWR International

Specifications

  • Coating:
    PTFE
  • Ø:
    9 mm (1/3")
  • Length:
    7.9 mm (5/16")
  • Description:
    VWR® Spinbar® Cuvette Stir Bar
  • Cat. no.:
    58949-030

Specifications

About this item

This magnetic stirring bar fits into the standard 10mm light path cuvette and provides rapid vertical and horizontal mixing with a minimum of vortexing when placed on a magnetic stirring machine.

  • One-piece molded unit will not scratch cuvettes; may also be used in test tubes
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon® PTFE low-friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclavable

Centrifugal pumping action, generated by the cross channels in the upper face, mixes without aeration. Photometric measurements can proceed uninterrupted while reagents are added.

Customers Who Bought This Also Bought