Rotors for Allegra® X-30 Series Centrifuges, Swinging-Bucket Microplate, Beckman Coulter®
Supplier: Beckman Coulter
Certificates
About this item
These rugged Beckman Coulter rotors are constructed from heavy-duty aluminum and are engineered to provide quiet, dependable service.
- Spins 2 x 3 microplates or 2 x 1 deep-well plates
- Applications include serial dilution of small, liquid volumes
- Unshielded
- Anodized aluminum material
- Quiet
Ordering information: Microplates not included. Rotors are supplied complete and ready to use. Bottles, wrenches, and accessories are available separately. Contact your Avantor representative for more information.
Specifications
- Catalog No:
- CABK361111
- CABKB01430
- Angle:
- 90°
- 90°