Benchtop Poison Cabinet, SciMatCo
Catalog #: 18500-064
Supplier: Scientific Materials
Specifications
- Depth:15¾"
- Width:16¾"
- Description:Acids and poisons bench cabinet
- Height:21¼"
- Cat. no.:18500-064
- Capacity:2x 1 gal. bottles
- Supplier No.:SC6080
Specifications
About this item
Constructed of 2.5 cm (1") multi-ply plywood, cabinet fits on laboratory bench, has one half depth fixed position shelf and a 5.1 cm (2") liquid-tight trough for spill containment.
- Door features a lock for safety, an easy-to-grip T-handle and a strong, continuous piano-type door hinge
- Liquid-tight trough retains hazardous spills
- Non-reactive wood
Poison cabinets are easily identified by gray color and large diamond shaped poison label.
Delivery: Supplied with internal metal lock and chemical resistant epoxy finish.
Customers Who Bought This Also Bought