To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:D4 Complete System
- Cat. no.:27373-562
- L×W×H:23×27×14 cm (9 × 105/8 × 51/2")
- Buffer Volume:1.8 L
- Supplier No.:D4
Specifications
About this item
This agarose gel electrophoresis system is designed to provide flat, even banding patterns and consistent results with hassle-free gel casting
No tape, grease, agarose seals, or other accessories are required. One gel tray (51 samples) or two stacked gel trays (102 samples) can be run in the buffer chamber while saving valuable bench space. Each of the two U.V. Transmissible (UVT) gel trays, measuring 16Wx17Lcm (65/16x611/16"), accommodates up to three combs, allowing the user to run up to three series of samples of equal distances. A stand-alone casting platform is included for casting two gels simultaneously. A single gel can be cast right in the buffer chamber.