To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:A2 Complete System
- Cat. no.:27373-554
- L×W×H:37.5×28×10 cm (143/4×11×315/16")
- Buffer Volume:2.3 L
- Supplier No.:A2-OK
Specifications
About this item
This system is ideal for teaching laboratories or for any laboratory that requires the ability to run multiple gels simultaneously
With this system, one teacher can instruct up to 12 students with only one power supply. This versatile system can run up to six 7Wx8Lcm (23/4x31/8") mini gels at one time. When used with double-sided combs, the system can screen up to 144 samples at once. The EasyCast* casting design allows for the casting of gels without tape or plugs. U.V. transmissible (UVT) gel trays may be placed directly on the transilluminator for photo documentation.