Specifications
- Unit dimensions:17.5W x 12.5D x 18H cm
- Description:Complete System
- Cat. no.:27373-566
- W×D×H:17.5 x 12.5 x 18 cm (67/8 x 47/8 x 71/8")
- Gel Dimensions:10W x 8L cm
- Supplier No.:P82
Specifications
About this item
For flat, even banding and crisp resolution
System includes an upper buffer chamber, lower buffer chamber, SuperSafe™ lid with attached power supply leads, two 1.3cm (1/2") thick wedges, and two 1.6 cm (5/8") thick wedges. The two sizes of wedges are provided to accommodate varying thickness of precast gels.
The system can run two 10 W x 10 L cm gels, or a single gel using the blocking plate. The system can accommodate 10 W x 8 L cm (4 x 31/8") gels utilizing the provided adapter. Gels can be poured or precast. The system is designed to allow gels to be easily placed into the chamber. A wedge is placed in front of the cassette to provide even pressure against the leakproof gasket and ensure that the cassette is held in the proper running position. A molded gasket provides a leak-resistant seal. The lid closes in one orientation only, ensuring that the positive and negative electrodes are not reversed. No cooling is required.