Owl™ Single Sided Vertical Electrophoresis System, Model P81, Thermo Scientific
Catalog #: 27373-431
Supplier: Thermo Fisher Scientific
Specifications
- Unit dimensions:18.4W x 8.3D x 12.7H cm
- Description:Complete System
- Cat. no.:27373-431
- Buffer Volume:150 ml
- Gel Dimensions:10L x 10W cm
- Supplier No.:P81
- Buffer volume:Upper Chamber: 100mL; Lower Chamber: 50mL
- Gel dimensions:10L x 10W cm (315/16 x 315/16")
Specifications
About this item
System features an extended upper buffer chamber that provides even heat distribution.
System may be used with precast gels, or with gels poured by hand.
Chamber brings the buffer into direct contact with the entire gel surface, providing efficient cooling. System requires less than 100 mL of buffer per run. Built-in, full length, spring-loaded side clamps attach to a molded gasket to ensure a leakproof seal without the risk of overtightening.
BodyTextAccessories: Combs are available in standard or preparative types.
Ordering information: Supplied with manufacturer’s three-year warranty. Complete system includes an upper and lower buffer chamber; SuperSafe™ lid with attached power supply leads; one blank glass plate; one notched glass plate; one notched alumina plate; two 10-well, 0.8 mm thick combs; and two 0.8 mm thick side spacers.
Customers Who Bought This Also Bought