To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Max. sample capacity:80 Samples
- Unit dimensions:30.5W x 15.2D x 7H cm (12 x 6 x 23/4")
- Description:MP1015 Horizontal Gel System
- Gel dimensions:10W x 15L cm (37/8 x 57/8")
- Cat. no.:IB53000
- Buffer Volume:500 ml
- Gel Dimensions:10 x 15 cm
- Supplier No.:53000
Specifications
About this item
Ideal for extended, high-resolution linear runs and economical quick screens
Buffer recirculation capability permits high resolution electrophoresis of nucleic acids in low ionic strength buffers. The casting fixture and outside unit design permit gel preparation while minimizing leaks. Well visualization stripes improve visibility and facilitate gel loading. The lid is vented to reduce condensation, and gel trays transmit 312nm UV light. An electrical gravity connector minimizes electrical hazards and eliminates sample disturbance during setup. Gold-plated electrical fittings and 99.9% platinum electrodes resist corrosion and withstand rapid, high-voltage polarity switching applications. The unit can be used with a sample quick loader. It accommodates up to four combs and is compatible with IBI MPH combs.