To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Max. sample capacity:30 Samples
- Unit dimensions:22.2W x 11.4D x 6.4H cm (83/4 x 41/2 x 21/2")
- Description:QS710 Horizontal Gel Unit
- Gel dimensions:7W x 10L cm (23/4 x 4")
- Cat. no.:IB51000
- Buffer Volume:250 ml
- Gel Dimensions:7 x 10 cm
- Supplier No.:51000
Specifications
About this item
Designed for quick screening and accurate, rapid separation of nucleic acids
Well visualization stripes improve visibility, facilitating gel loading. Vented lid reduces condensation, improving resolution of fast runs. Gel trays transmit 312nm UV light while allowing maximum sample capacity using multiple combs, or maximum running lengths using a single comb.
Electrical gravity connector minimizes electrical hazards and eliminates sample disturbance during setup. Safety-tipped power cords with retractable sleeves and plastic tips reduce electrical hazards.
Precision-made Delrin® analytical combs (sold separately) are available in a wide range of thicknesses and tooth configurations to accommodate multiple samples or large sample volumes.
Use for electrophoresis procedures that do not require buffer recirculation. Unit features corner leveling feet for perfectly straight gel runs, three-piece acrylic construction that resists warping, and a leveling bubble to ensure perfect gels. Three gel sizes allow applications ranging from quick screens to high-resolution separations.