E-LAB 2 Vacuum Pump, Edwards
Catalog #: CAA65-207-906
Supplier: Edwards
Specifications
- Description:Vacuum Pump
- Cat. no.:CAA65-207-906
- Supplier No.:A65-207-906
Specifications
About this item
This pump was specifically developed for wet chemistry and sample preparation
An internal cartridge design provides easy servicing and the continuous oil recycling permits constant running. The E-LAB 2 pump has a high efficiency mist filter which eliminates any oil smell.
It is ideal for use with rotary evaporators, centrifugal concentrators, vacuum ovens, distillation/extraction apparatus, fume hoods, and vacuum filtration apparatus.
Certifications: CSA certified and CE marked.
Ordering information: Supplied with manufacturer’s one-year warranty. Complete with a standard nozzle kit for connection to any system.
Customers Who Bought This Also Bought