VWR® Non-Return Valves
Catalog #: 46600-040
Supplier: VWR International
Specifications
- Description:Non-Return Valve, Crest 7.8/10.0 mm (5/16/13/32 ")
- Cat. no.:46600-040
- Nominal O.D.:8/9/10 mm (⁵⁄₁₆/¹¹⁄₃₂/¹³⁄₃₂")
Specifications
About this item
This polypropylene non-return valve protects tubing and vacuum systems from back flow when pressure falls in the system.
- Polypropylene construction
- Protects against back flow
The recommended working pressure range for this valve is 1.0 to 28.4psi.
Customers Who Bought This Also Bought