About this item
ClassMate™ laboratory hoods are designed to meet the needs of instructional laboratories.
The hood frame, spill trough, and standard hood air foil are constructed of white dry-powder epoxy-coated steel and aluminum. The low-profile spill trough is 3.2cm (11/4") deep and requires no cabinet overhang. The HOPEC IV air foil is a highly visible yellow. All hoods include pre-wired fluorescent lighting, ADA-compliant light and blower switches, removable interior cover plates for easy plumbing access, and a 20.3cm (8") diameter belled exhaust connection. A tissue screen below the exhaust outlet protects the remote blower from debris.
- High-performance, bypass airflow design
ClassMate™ laboratory hoods are designed to meet the needs of instructional laboratories.
Clear tempered safety glass provides excellent visibility even when hoods are placed back to back
Clear tempered safety glass provides excellent visibility even when hoods are placed back to back
The hood frame, spill trough, and standard hood air foil are constructed of glacier white powder-coated steel and aluminum. The low-profile spill trough is 3.2 cm (11/4”) deep and requires no cabinet overhang. All hoods include pre-wired LED lighting, ADA-compliant light and blower switches, removable interior cover plates for easy plumbing access, and a 12” (30.5 cm) dia. exhaust connection. A tissue screen below the exhaust outlet protects the remote blower from debris.
High-performance, 3-piece baffle pivots for cleaning. The glass front is 371/2” (95.25 cm) high, 0.6 cm (1/4”) thick, and at a 5° angle. The vertical-rising sash of the combination style hoods also include horizontal-sliding panels.
All hoods feature a high-performance, bypass airflow design. Baffles direct airflow to the rear of the interior. A pivoting air foil features a trough for secondary spill containment. Models with a combination style sash, which adds energy-saving and ergonomic benefits, are available.
Specifications
- Catalog No:
- 76045-640
- 76045-638
- 76045-636
- 76045-634
- 76045-632
- 76045-630
- 76045-628
- 76045-922
- 76045-646
- 76045-644
- 76045-642
- 76045-624
- 76045-622
- 76045-620
- 76045-618
- 76045-616
- 76045-910
- 76045-908
- 76045-656
- 76045-906
- 76045-654
- 76045-652
- 76045-650
- 76045-626
- 76045-648
- 76045-920
- 76045-918
- 76045-916
- 76045-914
- 76045-912
- Environmentally Preferable:
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Outside depth:
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- 80.1 cm (32.7")
- Outside height:
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- 149.9 cm (59")
- Vertical sash opening:
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- 71.1 cm (28") max.
- Environmental certification:
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood
- It meets the SEFA definition of a High Performance Fume hood