Order Entry
ContactUsLinkComponent
Minimate™ EVO Tangential Flow Filtration System and Capsules, Cytiva (Formerly Pall Lab)
Minimate™ EVO Tangential Flow Filtration System and Capsules, Cytiva (Formerly Pall Lab)
Catalog #: CA76409-724
Supplier:  Cytiva
Minimate* EVO tangential flow system, Type: TFF Cassettes and Modules, Reservoir Material: Polysulfone, Volume: < 500 mL, Dimensions30.7 W x 48.2 D x 20.8 H cm, For Tangential Flow Filtration, Universal plug, Complete system with Pump and Reservoir
Minimate™ EVO Tangential Flow Filtration System and Capsules, Cytiva (Formerly Pall Lab)
Catalog #: CA76409-724
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    EVO tangential flow filtration system
  • Operating temperature range:
    0...50 °C (0...106 °F)
  • Effective filtration area:
    50 cm2
  • Max. operating pressure:
    4.1 bar (410 kPa, 60 psi)
  • Dimensions:
    307×482×208 mm (12.1×19×8.2")
  • Cat. no.:
    CA76409-724
  • Electrical:
    110 - 230 VAC

Specifications

About this item

Reusable TFF devices and system for bioprocessing applications accelerates and simplifies processing of up to 1 L.

  • High product recovery
  • Minimal sample loss
  • Greater efficiency
  • Gentle processing
  • Plug-and-play processing
  • Scalable operation
  • Recommended crossflow: 30 - 80 ml/min.

Versatile, self-contained capsules include all fittings and tubing to simplify installation into filtration setups and will work with any laboratory pump. Dual pressure gauges allow trans-membrane pressure (TMP) differential calculations.

Capsules feature glass-reinforced polypropylene housing, polypropylene screens, housing sealing with fittings, polyethylene membrane plate/filtrate channels, and an internal gasket made of ethylene propylene elastomer.

The Minimate™ TFF system, designed to work with the Minimate™ TFF capsule, is ideal for a variety of tangential flow filtration applications in biotechnology, molecular biology, protein chemistry, microbiology, and immunology. This ready-to-use system can concentrate and desalt proteins, peptides or nucleic acid sample volumes of up to 1 L or more and efficiently concentrate samples to as little as 5 ml. System also features gentle and efficient processing, easy cleaning and maintenance, compact design, and high concentration factors.

The optimized flow path design of the capsules coupled with low protein binding Omega™ (modified polyethersulfone) ultrafiltration membrane minimize non-specific binding. Capsules allow concentration and desalting processes to be performed on the same system with minimal user intervention. Several Minimate™ TFF capsules may be connected in parallel for increased membrane area.

Certifications: Each Minimate capsule is 100% integrity tested during manufacture to ensure reliable performance. For critical applications, the user can re-test the integrity after initial use. A Certificate of Quality is included with each capsule.
BodyTextAccessories: Minimate Fittings Kit (29301-928) contains: Male luer to 3.2 mm (¹/₈") hose barb, Female luer to 3.2 mm (¹/₈") hose barb, 3.2 mm (¹/₈") Int.Ø tubing, tubing screw clamp, tubing clamps, adhesive strips (loop and hook).
Ordering information: System includes a pump, pump head, 2 pressure gauges, 500 ml reservoir, stir plate and magnetic stir bar, valves, fittings, and a compact drip tray. 110 to 230 VAC input.
Customers Who Bought This Also Bought