VWR® Qualitative Fluted Circles, Folded, Grade 313
Supplier: VWR International
CE Compliant
Certificates
About this item
Used for drugs, pharmaceuticals, liquors, tinctures, chromatography, and protein-free filtrations in the clinical laboratory.
- Medium fine porosity
- Medium flow rate
- Assorted sizes
Fluted circles are available in a variety of sizes and boast a medium flow rate.
Specifications
- Catalog No:
- CA28333-123
- CA28333-087
- CA28333-065
- CA28333-043
- CA28333-021
- Particle retention:
- 5 µm
- 5 µm
- 5 µm
- 5 µm
- 5 µm
- Porosity:
- Medium Fine
- Medium fine
- Medium fine
- Medium fine
- Medium fine
- Flow rate:
- Medium
- Medium
- Medium
- Medium
- Medium
Specifications
Related Information
Certificates
Search by Lot Number28333-065_A27482274
Conformity declaration
F&BFOCUS Volume 5 Issue 1w VWR F&BFOCUS
Product Literature
Customers Who Bought This Also Bought