VWR® Qualitative Crepe Filter Paper, Grade 417
Supplier: VWR International
CE Compliant
Certificates
About this item
Good for rapid filtrations where fine particle retention is not required.
- Coarse porosity
- Fast flow rate
- Assorted sizes
Specifications
- Catalog No:
- CA28313-240
- CA28313-206
- CA28313-181
- CA28313-160
- CA28313-148
- CA28313-126
- CA28313-104
- CA28313-080
- CA28297-868
- CA28313-068
- CA28313-057
- CA28313-046
- Particle retention:
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- Porosity:
- Coarse
- Coarse
- Coarse
- Coarse
- Coarse
- Coarse
- Coarse
- Coarse
- Coarse
- Coarse
- Coarse
- Coarse
Specifications
Related Information
Certificates
Search by Lot NumberCoQ VWR® Grade 417 Filter Paper, Qualitative, Crepe.pdf
Conformity declaration
Customers Who Bought This Also Bought