Nalgene® Filter Holder with Funnel, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Polysulfone filter holder with funnel can be used with any filtering flask or manifold that accepts a suitable stopper with hole.
Note: For lab use only. Not for in vitro diagnosis or parenterals.
Autoclavable.
Ordering information: Unit comes complete with analytical filter support, sterilization filter support, and vacuum gasket, which takes the place of a No.7 or 8 rubber stopper with 14.3 mm (9/16") hole.
Specifications
- Catalog No:
- 28199-437
- 28199-441
- Filtration area:
- 13.3 cm²
- 13.3 cm²