Anti-WFDC2 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: WFDC2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77706-556
- 77706-555
- Antigen symbol:
- WFDC2
- WFDC2
- Antigen synonyms:
- Epididymis specific whey acidic protein type four disulfide core|epididymal protein 4|WFDC2_HUMAN|WFDC2|WAP four disulfide core domain 2|WFDC 2|Major epididymis specific protein E4|EDDM4|WAP four-disulfide core domain protein 2|WAP5|WAP 5|MGC57529|WAP four disulfide core domain protein 2|Major epididymis-specific protein E4|WAP domain containing protein HE4|Putative protease inhibitor WAP5|dJ461P17.6|WAP domain containing protein HE4 V4|HE 4|Epididymal secretory protein E4
- Epididymis specific whey acidic protein type four disulfide core|epididymal protein 4|WFDC2_HUMAN|WFDC2|WAP four disulfide core domain 2|WFDC 2|Major epididymis specific protein E4|EDDM4|WAP four-disulfide core domain protein 2|WAP5|WAP 5|MGC57529|WAP four disulfide core domain protein 2|Major epididymis-specific protein E4|WAP domain containing protein HE4|Putative protease inhibitor WAP5|dJ461P17.6|WAP domain containing protein HE4 V4|HE 4|Epididymal secretory protein E4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14508
- Q14508
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human WAP four-disulfide core domain protein 2 protein (31-124AA)
- Recombinant Human WAP four-disulfide core domain protein 2 protein (31-124AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together