Anti-UBE2K Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBE2K
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77706-425
- 77706-424
- Antigen symbol:
- UBE2K
- UBE2K
- Antigen synonyms:
- ubiquitin conjugating enzyme E2K (UBC1 homolog, yeast)|HYPG|Ubiquitin conjugating enzyme E2 25 kDa|Huntingtin-interacting protein 2|E2 25K|UBE2K_HUMAN|UBC1|Ubiquitin carrier protein|Ubiquitin-conjugating enzyme E2(25K)|Huntingtin interacting protein 2|E2(25K)|UBE2K|LIG|Ubiquitin protein ligase|Ubiquitin-protein ligase|ubiquitin conjugating enzyme E2K|HIP-2|Ubiquitin-conjugating enzyme E2-25K|Ubiquitin-conjugating enzyme E2-25 kDa|Ubiquitin-conjugating enzyme E2 K
- ubiquitin conjugating enzyme E2K (UBC1 homolog, yeast)|HYPG|Ubiquitin conjugating enzyme E2 25 kDa|Huntingtin-interacting protein 2|E2 25K|UBE2K_HUMAN|UBC1|Ubiquitin carrier protein|Ubiquitin-conjugating enzyme E2(25K)|Huntingtin interacting protein 2|E2(25K)|UBE2K|LIG|Ubiquitin protein ligase|Ubiquitin-protein ligase|ubiquitin conjugating enzyme E2K|HIP-2|Ubiquitin-conjugating enzyme E2-25K|Ubiquitin-conjugating enzyme E2-25 kDa|Ubiquitin-conjugating enzyme E2 K
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61086
- P61086
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ubiquitin-conjugating enzyme E2 K protein (2-200AA)
- Recombinant Human Ubiquitin-conjugating enzyme E2 K protein (2-200AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together