Anti-MDK Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: MDK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77710-412
- 77710-413
- Antigen synonyms:
- Amphiregulin-associated protein|ARAP|MK_HUMAN|Amphiregulin associated protein|Neurite outgrowth-promoting protein|Retinoic acid inducible factor|NEGF2|MK1|NEGF 2|Neurite growth promoting factor 2|Midkine|Mdk|Midgestation and kidney protein|FLJ27379|MK 1|Neurite outgrowth-promoting factor 2|MK|Neurite outgrowth promoting protein
- Amphiregulin-associated protein|ARAP|MK_HUMAN|Amphiregulin associated protein|Neurite outgrowth-promoting protein|Retinoic acid inducible factor|NEGF2|MK1|NEGF 2|Neurite growth promoting factor 2|Midkine|Mdk|Midgestation and kidney protein|FLJ27379|MK 1|Neurite outgrowth-promoting factor 2|MK|Neurite outgrowth promoting protein
- Gene ID:
- P21741
- P21741
- Immunogen:
- Fusion protein of Human MDK
- Fusion protein of Human MDK
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together