Anti-CASP8AP2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:25-1:100
Type:
Antigen: CASP8AP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77710-156
- 77710-157
- Antigen synonyms:
- Caspase 8 associated protein 2|AA387232|RIP25|C8AP2_HUMAN|OTTMUSP00000005125|KIAA1315|FLASH homolog RIP 25|CASP 8 associated protein 2|FLICE associated huge|FLICE associated huge protein|CED4|RP23-69G24.1|CASP8-associated protein 2|CED 4|FLICE-associated huge protein|CASP8AP2|FLASH homolog|RIP 25|D4Ertd659e|CASP8 associated protein 2|mKIAA1315|FLJ11208|FLASH homolog RIP25
- Caspase 8 associated protein 2|AA387232|RIP25|C8AP2_HUMAN|OTTMUSP00000005125|KIAA1315|FLASH homolog RIP 25|CASP 8 associated protein 2|FLICE associated huge|FLICE associated huge protein|CED4|RP23-69G24.1|CASP8-associated protein 2|CED 4|FLICE-associated huge protein|CASP8AP2|FLASH homolog|RIP 25|D4Ertd659e|CASP8 associated protein 2|mKIAA1315|FLJ11208|FLASH homolog RIP25
- Gene ID:
- Q9UKL3
- Q9UKL3
- Immunogen:
- Synthetic peptide of Human CASP8AP2
- Synthetic peptide of Human CASP8AP2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together