Anti-CARD8 Rabbit Polyclonal Antibody
New Product
ELISA:1:1000-1:2000, IHC:1:25-1:100
Type:
Antigen: CARD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77710-139
- 77710-138
- Antigen synonyms:
- CARDINAL|Dakar|FLJ18119|Caspase recruitment domain protein 8|CARD8_HUMAN|Tumor up-regulated CARD-containing antagonist of CASP9|CARD-inhibitor of NF-kappa-B-activating ligand|Caspase recruitment domain-containing protein 8|CARD 8|Caspase recruitment domain family member 8|NDPP 1|KIAA0955|tumor up-regulated CARD-containing antagonist of caspase nine|Apoptotic protein NDPP 1|Tumor upregulated CARD containing antagonist of CASP9|DKFZp779L0366|TUCAN|Tumor up regulated CARD containing antagonist of caspase 9|Tumor up regulated CARD containing antagonist of CASP9|FLJ18121|MGC57162|CARD inhibitor of NF kappaB activating ligands|CARD inhibitor of NF kappa B activating ligand|CARD inhibitor of NF kappaB activating ligand|DACAR|NDPP|Caspase recruitment domain containing protein 8|NDPP1|Apoptotic protein NDPP1|CARD8
- CARDINAL|Dakar|FLJ18119|Caspase recruitment domain protein 8|CARD8_HUMAN|Tumor up-regulated CARD-containing antagonist of CASP9|CARD-inhibitor of NF-kappa-B-activating ligand|Caspase recruitment domain-containing protein 8|CARD 8|Caspase recruitment domain family member 8|NDPP 1|KIAA0955|tumor up-regulated CARD-containing antagonist of caspase nine|Apoptotic protein NDPP 1|Tumor upregulated CARD containing antagonist of CASP9|DKFZp779L0366|TUCAN|Tumor up regulated CARD containing antagonist of caspase 9|Tumor up regulated CARD containing antagonist of CASP9|FLJ18121|MGC57162|CARD inhibitor of NF kappaB activating ligands|CARD inhibitor of NF kappa B activating ligand|CARD inhibitor of NF kappaB activating ligand|DACAR|NDPP|Caspase recruitment domain containing protein 8|NDPP1|Apoptotic protein NDPP1|CARD8
- Gene ID:
- Q9Y2G2
- Q9Y2G2
- Immunogen:
- Fusion protein of Human CARD8
- Fusion protein of Human CARD8
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together