Order Entry
Canada
ContactUsLinkComponent
 
Anti-THAP11 Rabbit Polyclonal Antibody (Biotin)
New Product
 
Anti-THAP11 Rabbit Polyclonal Antibody (Biotin)
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Catalog No:
  • 77723-791
  • 77723-790
  • Antigen symbol:
  • THAP11
  • THAP11
  • Antigen synonyms:
  • CTG B43a|THAP domain containing 11|CTG B45d|RONIN|THA11_HUMAN|THAP 11|THAP domain-containing protein 11|Thap11|HRIHFB2206
  • CTG B43a|THAP domain containing 11|CTG B45d|RONIN|THA11_HUMAN|THAP 11|THAP domain-containing protein 11|Thap11|HRIHFB2206
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q96EK4
  • Q96EK4
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human THAP domain-containing protein 11 protein (1-313AA)
  • Recombinant Human THAP domain-containing protein 11 protein (1-313AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on Blue Ice
  • Shipped on Blue Ice

 

Frequently Bought Together