Anti-APOBEC3D Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: APOBEC3D
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77723-698
- 77723-699
- Antigen symbol:
- APOBEC3D
- APOBEC3D
- Antigen synonyms:
- isoform CRA_b|hCG_41704|Apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3E pseudogene|APOBEC3E|DNA dC >dU editing enzyme APOBEC 3D|bK150C2.9|HCG41704|Apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3D|APOBEC3DE|ABC3D_HUMAN|BK150C2.9 protein|Probable DNA dC->dU-editing enzyme APOBEC-3D|DNA dC->dU-editing enzyme APOBEC-3D|ARP6|A3D|APOBEC3D
- isoform CRA_b|hCG_41704|Apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3E pseudogene|APOBEC3E|DNA dC >dU editing enzyme APOBEC 3D|bK150C2.9|HCG41704|Apolipoprotein B mRNA editing enzyme catalytic polypeptide like 3D|APOBEC3DE|ABC3D_HUMAN|BK150C2.9 protein|Probable DNA dC->dU-editing enzyme APOBEC-3D|DNA dC->dU-editing enzyme APOBEC-3D|ARP6|A3D|APOBEC3D
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96AK3
- Q96AK3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human DNA dC->dU-editing enzyme APOBEC-3D protein (250-371AA)
- Recombinant Human DNA dC->dU-editing enzyme APOBEC-3D protein (250-371AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue Ice
- Shipped on Blue Ice
Specifications
Frequently Bought Together