Order Entry
Canada
ContactUsLinkComponent
Anti-ATP6V0A1 Rabbit Polyclonal Antibody
Anti-ATP6V0A1 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
Anti-ATP6V0A1 Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77723-638
  • 77690-353
  • Antigen symbol:
  • ATP6V0A1
  • ATP6V0A1
  • Antigen synonyms:
  • V-type proton ATPase 116 kDa subunit a isoform 1|Vacuolar proton pump subunit 1|V ATPase 116 kDa|V ATPase 116 kDa isoform a1|H(+) transporting two sector ATPase 116 kDa accessory protein A1|ATPase H+ transporting lysosomal V0 subunit a1|ATPase H+ transporting lysosomal non catalytic accessory protein 1 (110/116kD)|V type proton ATPase 116 kDa subunit a|Clathrin coated vesicle/synaptic vesicle proton pump 116 kDa subunit|VPP1|Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit|ATPase H+ transporting lysosomal (vacuolar proton pump) non catalytic accessory protein 1A (110/116kD)|V-ATPase 116 kDa isoform a1|ATP6V0A1|a1|Vacuolar type H(+) ATPase 115 kDa subunit|Stv1|V type proton ATPase 116 kDa subunit a isoform 1|Vacuolar proton pump, subunit 1|ATP6N1A|VPP1_HUMAN|Vacuolar proton translocating ATPase 116 kDa subunit A|Vacuolar proton translocating ATPase 116 kDa subunit a isoform 1|Vph1|ATPase H+ transporting V0 subunit a1|DKFZp781J1951|Vacuolar adenosine triphosphatase subunit Ac116|ATP6N1|ATPase, H+ transporting, lysosomal, noncatalytic accessory protein 1A
  • V-type proton ATPase 116 kDa subunit a isoform 1|Vacuolar proton pump subunit 1|V ATPase 116 kDa|V ATPase 116 kDa isoform a1|H(+) transporting two sector ATPase 116 kDa accessory protein A1|ATPase H+ transporting lysosomal V0 subunit a1|ATPase H+ transporting lysosomal non catalytic accessory protein 1 (110/116kD)|V type proton ATPase 116 kDa subunit a|Clathrin coated vesicle/synaptic vesicle proton pump 116 kDa subunit|VPP1|Clathrin-coated vesicle/synaptic vesicle proton pump 116 kDa subunit|ATPase H+ transporting lysosomal (vacuolar proton pump) non catalytic accessory protein 1A (110/116kD)|V-ATPase 116 kDa isoform a1|ATP6V0A1|a1|Vacuolar type H(+) ATPase 115 kDa subunit|Stv1|V type proton ATPase 116 kDa subunit a isoform 1|Vacuolar proton pump, subunit 1|ATP6N1A|VPP1_HUMAN|Vacuolar proton translocating ATPase 116 kDa subunit A|Vacuolar proton translocating ATPase 116 kDa subunit a isoform 1|Vph1|ATPase H+ transporting V0 subunit a1|DKFZp781J1951|Vacuolar adenosine triphosphatase subunit Ac116|ATP6N1|ATPase, H+ transporting, lysosomal, noncatalytic accessory protein 1A
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q93050
  • Q93050
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human V-type proton ATPase 116 kDa subunit a isoform 1 protein (80-260AA)
  • Recombinant Human V-type proton ATPase 116 kDa subunit a isoform 1 protein (80-260AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on Blue Ice
  • Shipped on Blue Ice

Specifications

Frequently Bought Together